Potassium hydroxide 0.5 mol/l (0.5 N) in ethanol, Titripur® Reag. Ph. Eur., Reag. USP volumetric solution, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Potash caustic, Caustic potash

M-Clarity™ quality level = MQ100

1.09114.1000 1.09114.1000DP 1.09114.1003
1.09114.1000EA 74.9 GBP
1.09114.1000 MERC1.09114.1000DP 1.09114.1003
Potassium hydroxide 0.5 mol/l (0.5 N) in ethanol, Titripur® Reag. Ph. Eur., Reag. USP volumetric solution, Supelco®
Potassium hydroxide

Merck have introduced into the range of Titripur® Titrants some packs with additional technology - Supelco® SmartTitrants that use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. MettlerToledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tag.

Formula: KOH
Melting Pt: –114.5 °C (EtOH)
Density: 0.85 g/cm³ (20 °C)
Flash Pt: 12 °C (EtOH)
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 2924
ADR: 3 (8),II

Order Now

Specification Test Results

Form liquid
Amount-of-substance concentration 0.4975 - 0.5025
Measurement uncertainty +/- 0.0015
Traceability NIST SRM


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR