Humidity chambers, HCP series
Supplier: MEMMERT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
HCP50 HCP105 HCP150 HCP240
MEMMHCP50EA 7530 GBP
MEMMHCP50 MEMMHCP105 MEMMHCP150 MEMMHCP240
Humidity chambers, HCP series
Testing Chambers Humidity Chambers
Humidity chambers which have corrosion resistant, stainless steel inner chamber and housing for long-term climatic tests at constant temperature and humidity. Ideal for food processing, cosmetics, pharmaceutical, biotechnology and electronics industries. Units feature an auto diagnostic system with fault indication for temperature and humidity control. The working chamber is heated on all six sides; this together with electronic humidity control prevents condensation forming in the chamber. Distilled water is supplied from an external 2,5 litre tank by means of a self priming pump.More Product Information