Transport swabs, Transwab®
Supplier: MEDICAL WIRE EQUIPMENT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
MW177S MW176S MW176SE2
MEDWMW177SEA 748 GBP
MEDWMW177S MEDWMW176S MEDWMW176SE2
Transport swabs, Transwab®
Swabs Transport Swabs
Transwab with a narrow shaft and bud for nasopharyngeal or urethral sampling.
  • Nasopharyngeal sampling such as for Bordetella pertussis
  • Open cell flow-through allows full access and release of sampled organisms
  • Optimum sample pick-up and release
Used in conventional and automated processing. Compatible with automated systems.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904