Staining tiles
Supplier: MARIENFELD
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1405500 1406405 1406703 1406809 1406506 1406708
MARI1405500EA 2190 GBP
MARI1405500 MARI1406405 MARI1406703 MARI1406809 MARI1406506 MARI1406708
Staining tiles
Staining Tiles
  • Bevelled edges
  • Ground and polished cavities
  • Clipped corners: Except the one with twelve wells