Adhesion slides, HistoBond®+
Supplier: MARIENFELD
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
0810401 0810411 0810421 0810431 0810441 0810451
MARI0810401EA 52.6 GBP
MARI0810401 MARI0810411 MARI0810421 MARI0810431 MARI0810441 MARI0810451
Adhesion slides, HistoBond®+
Slides Adhesive Slides
These adhesion slides are composed of silane treated surfaces and 90° ground edges.
  • Coloured marking areas on one end
  • Specially used for applications of colour coding
  • Improved identification of specimens
Certifications: IVD directive 98/79/EC