pH indicator papers, Duotest and Tritest
Supplier: MACHEREY-NAGEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
90301 90302 90303 90304 90311 90313 90501 90502 90510 90511 90305 90306 90312 90314 90315 90316
662-0235EA 16.6 GBP
662-0235 MANA90302 516-6011 MANA90304 MANA90311 MANA90313 662-0195 662-0203 662-0115 662-0116 MANA90305 MANA90306 MANA90312 MANA90314 MANA90315 MANA90316
pH indicator papers, Duotest and Tritest
Test Papers pH Indicator Papers and Test Strips
By the combination of several indicators on one test paper, a better recognition between different pH values is achieved. This makes the correct pH reading easier.
  • Long term stable and protected against external influences due to plastic reel
  • No accessories needed, always ready to use
  • Reliable results due to easy and accurate read-off from colour scale
  • Several indicators for more precise readings
Duotest indicator papers combine two different indicator zones on a single strip. The zones are separated by a hydrophobic barrier, which effectivelyMore Product Information