Crimp neck vials, N 11
Supplier: MACHEREY-NAGEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
70201CG 70201HP 702141 70214CG 702885 702888 702891 702892 702014 702015 702016 70201HP.2 702075 702076 702101HP 702859 702867 702882
MANA70201CGEA 14.7 GBP
MANA70201CG MANA70201HP MANA702141 MANA70214CG MANA702885 MANA702888 MANA702891 MANA702892 MANA702014 MANA702015 MANA702016 MANA70201HP.2 MANA702075 MANA702076 MANA702101HP MANA702859 MANA702867 MANA702882
Crimp neck vials, N 11
Vials
This broad range of crimp neck vials is available with small or wide opening.
  • Made of clear and amber glass or transparent polypropylene
  • Silanised glass vials for highest demands
  • Flat bottom
  • Label and scale for convenient handling
  • Different inserts available
  • State-of-the-art micro-vials in different versions for small sample volumes
Useable without adapter for the common autosamplers of Agilent, CTC, Dionex (Thermo Scientific), PerkinElmer, Shimadzu, Thermo Scientific, Varian (Agilent), VWR® (Merck / Hitachi), Waters etc.
Ready-to-use crimp capsMore Product Information