Filter paper for breweries
Supplier: MACHEREY-NAGEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
410015 427018 527012 527015 527018 527024 527032 562024 562050 562015 410005 410007 410009 410011 410012 410018 410024 427005 427007 427009 427011 427012 427015 427024 427032 441009 441011 441012 441015 441018 441024 441032 527005 527007 527009 527011 541009 541011 541012 541015 541018 541024 541032 562018 562027 562032 562038 562040 562045
MANA410015EA 22.2 GBP
MANA410015 MANA427018 MANA527012 MANA527015 MANA527018 515-9760 515-9761 MANA562024 MANA562050 MANA562015 MANA410005 MANA410007 MANA410009 MANA410011 MANA410012 MANA410018 MANA410024 MANA427005 MANA427007 MANA427009 MANA427011 MANA427012 MANA427015 MANA427024 MANA427032 MANA441009 MANA441011 MANA441012 MANA441015 MANA441018 MANA441024 MANA441032 MANA527005 MANA527007 MANA527009 MANA527011 MANA541009 MANA541011 MANA541012 MANA541015 MANA541018 MANA541024 MANA541032 MANA562018 MANA562027 MANA562032 MANA562038 MANA562040 MANA562045
Filter paper for breweries
Filters Filter Papers Qualitative Filter Paper
Filter papers used in beverage industries. Available in plain and folded form.
Grade MN 620: Used to efficiently decarbonate beer.
Grade MN 321 (nitrogen-free) and 614: Used for malt analysis.
Grade MN 960: Used in beverage industries for filtration of wines, fruit juices.