Lens tissue papers, MN 13
Supplier: MACHEREY-NAGEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
418101 418102 118000
MANA418101EA 33.3 GBP
MANA418101 MANA418102 MANA118000
Lens tissue papers, MN 13
Wipes Disposable Wipes and Towels Lens Papers
Very thin and soft filtration paper for cleaning optical glasses and cuvettes.