Guanidinium hydrochloride 8 mol/l in aqueous solution, Biotechnology Grade

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Guanadine hydrochloride, Aminoformamidine hydrochloride, Aminomethanamidine hydrochloride, Guanidine HCI, Guanidine chloride, Guanidinium chloride, Guanidine hydrochloride

M113-4LEA 1910 GBP
M113-4L
Guanidinium hydrochloride 8 mol/l in aqueous solution, Biotechnology Grade
Guanidinium hydrochloride

A concentrated solution prepared with Biotechnology Grade powder for convenient use in nucleic acid and protein purification procedures.

Formula: NH₂C(=NH)NH₂·HCl
MW: 95.53 g/mol
Melting Pt: 185
Storage Temperature: Ambient
MDL Number: MFCD00013026
CAS Number: 50-01-1
EINECS: 200-002-3
Merck Index: 13,04578
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da5a871cab977e6d9dc666b457732eb1e869a714&vid=b0d4fcb044b6106fce8e40448dbf2ce6e7077ad7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Protease none detected


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR