Caps for test/culture tubes without rim
Supplier: LUDI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
184010631 184010741 184010742 184010743 184010744 184010745
LUDI184010631EA 328 GBP
LUDI184010631 LUDI184010741 LUDI184010742 LUDI184010743 LUDI184010744 LUDI184010745
Caps for test/culture tubes without rim
Closures Test/Sample Tube Closures
Made of anodised aluminium.
  • Springs clamp against the outer wall of the vessel
  • Anodised and coloured
  • Autoclavable
Without grip. Cap for sterile but not hermetic closing of test tubes and other vessels.