WHEATON® Dropping Bottle Caps, 15-415
Supplier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
W001637RB
LSUKW001637RBEA 0 GBP
LSUKW001637RB
WHEATON® Dropping Bottle Caps, 15-415
Closures Dropper Caps
Polypropylene dropping bottle caps for use in conjunction with WHEATON® dropping bottles and tips to provide a complete solution.
  • Manufactured from polypropylene
  • Good chemical resistance
  • Autoclavable
Caps conform to stringent ISO and quality standards, and are lot controlled. Double bagged and twist tied to help reduce particulates.