Turbidity Standards and Sets for Hach® Turbidimeter
Supplier: Lovibond
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
48011050 48012050 48011150 48012950 48012912 48012312 48019712 48011512 48012012
LOVI48011050EA 131 GBP
LOVI48011050 LOVI48012050 LOVI48011150 LOVI48012950 LOVI48012912 LOVI48012312 LOVI48019712 LOVI48011512 LOVI48012012
Turbidity Standards and Sets for Hach® Turbidimeter
Standards Turbidity Standards
The T-CAL® sets with formazine based, long time stable primary standards are offered in different concentrations, fitting perfectly to your instrument for calibration and verification.
  • Suitable for Hach® instruments
  • Stabilised solution with long shelf life
  • Prevention of measurement errors due to accurate mixing
  • Save time and eliminate errors - no need for dilution
  • Avoid operator liability related to improper dilutions
  • Use continually for up to 24 months from production
T-CAL® Turbidity standards are stable formazin standards prepared in ready-to-use concentrations to ensure accurate turbidity measurements. All concentrations are verified to meet tight quality control specifications, are EPA and ISO compliant for reporting purposes and guaranteed to save you time. As a primary standard, T-CAL® Turbidity standards can be used to calibrate or verify any manufacturer’s turbidimeter.
Certifications: Conform to USEPA 180.1 and ISO 7027 norms.