Inoculating loops, PS
Supplier: Local
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
A3 A4 A4G B3 B4 B4G C3 C4 C4G
612-5695EA 29.1 GBP
612-5695 612-5769 LOCAA4G 612-5694 612-5768 LOCAB4G 612-5683 612-5767 LOCAC4G
Inoculating loops, PS
Spreaders Inoculating Loops and Needles
Calibrated disposable PS inoculating loops for different applications and microbiological methods.
  • Loops have an ultra-smooth surface for easy streaking
  • Eliminates the need for flaming
  • Aseptically manufactured in cleanroom giving a sterility assurance level 10⁻³