LiChrospher® RP-18E, HPLC Columns
Supplier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
LISPRP18E-5-250AEA 748 GBP
LISPRP18E-5-250A LISPRP18E5250F LISPRP18E-5-10C5 LISPRP18E-5-125A LISPRP18E5125F LISPRP18E5125S LISPRP18E-5-150A LISPRP18E5150F LISPRP18E5150M LISPRP18E5150S LISPRP18E5250S LISPRP18E-5-50AF LISPRP18E5200A LISPRP18E5300A LISPRP18E10250 SPRP18E510C5SK LISPRP18E5250P
LiChrospher® RP-18E, HPLC Columns
Chromatography Columns
These LiChrospher® columns are silica available both end capped and non-end capped C18 phases. This feature enhances the chromatography selectivity options.
  • Spherical porous silica
  • High efficiency packed columns