LiChrospher®, HPLC Columns
Supplier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
LISP100-5-50AEA 0 GBP
LISP100-5-50A LISP60-5-150A LISP60-5-250A LISP60-5-50AS LISPCN-5-125A LISPCN-5-250A LISPNH-5-100A LISPNH-5-125A LISPNH-5-250A LISPOH-5-125A LISPOH-5-150A LISPOH-5-250A LISPOH-5-50AM LISP100-5-150A LISP100-5-250A LISP60-5-10C5 LISP60-5-125AF LISP60-5-150AM LISP60-5-150AS LISP60-5-250AF LISP60-5-250AM LISPCN-5-125AF HICHLISP100-5100AF HICHLISP100-5100AM HICHLISP100-5100AS LISP100-5-10C5 LISPCN-5-250AF LISPNH-5-10C5 LISPNH-5-125AF LISPNH-5-250AF LISPOH-5-100AF LISPOH-5-100AM LISPOH-5-10C5 LISPOH-5-10CM5 LISPOH-5-125AF LISPOH-5-150AM LISPOH-5-150AS LISPOH-5-250AF LISPOH-5-250AM LISPOH-5-250AS LISP60-5-10CP3 LISP60-5-250P LISPOH510C5SK LISPOH-5-10CE5 LISPOH-5-150M LISPOH-5-250P LISPCN-5-75AF
LiChrospher®, HPLC Columns
Chromatography Columns
LiChrospher® is a silica available in both endcapped and non-endcapped C8 and C18 phases.
  • Spherical porous silica
  • Base deactivated LiChrospher® RP-select B
  • High efficiency packed columns