Swabs for monitoring of surface, Contam
Supplier: LIOFILCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
86102 86100 86103 86101 86105 86104
86102.EA 158 GBP
86102. 86100. 86103. 86101. 86105. LIOF86104
Swabs for monitoring of surface, Contam
Monitoring Systems Hygiene Monitoring Systems
For microbial monitoring of pathogens on surfaces.
  • Easy to use
  • Clear colour changes
  • No additional reagents
  • Results in 18/24 hours
  • 10 CFU/ml detection limit