Buffered NaCl Peptone Solution pH 7,0
Supplier: LIOFILCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
402730 403410
LIOF402730EA 44.2 GBP
LIOF402730 LIOF403410
Buffered NaCl Peptone Solution pH 7,0
Buffers Cell Culture Buffers
Buffered NaCl Peptone Solution pH 7,0 is used for dissolving, suspending and diluting test samples. Its composition conforms to recommendations in the Harmonised Pharmacopoeias for the microbiological examination of non-sterile products.
Certifications: This solution also conforms to ISO 21149 for the detection of viable microorganisms in cosmetics.
Ordering information: For professional use only.