Performance Professional Refrigerators with Dynamic cooling and Plastic Inner Liner, FRFvg
Supplier: Liebherr
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
FRFVG5501 FRFVG4001 FRFVG6501
LIEBFRFVG5501EA 1989 GBP
LIEBFRFVG5501 LIEBFRFVG4001 LIEBFRFVG6501
Performance Professional Refrigerators with Dynamic cooling and Plastic Inner Liner, FRFvg
Refrigerators
Refrigerators designed for professional use. Reliably store non temperature-sensitive items even at ambient temperatures up to +40 °C with a constant cooling power. They are enormously robust, long-lasting, easy to clean and particularly sustainable, not only thanks to natural refrigerant. At the same time, they fulfil all the valid directives, standards and norms, including in terms of hygiene, HACCP concept and GN containers. The efficient cooling system not only ensures low energy and overall operating cost, also safe storage of items not requiring validation of temperature stability andMore Product Information