Overhead stirrer tools, anchor stirrers
Supplier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
06320628 06320637 06320649 06320658 06320670 06321049 06321058 06321070 06321073 06321076 06321079 06321085 06321088 06321094 06321573 06321576 06321579 06321588 06321594
LENZ06320628EA 245 GBP
LENZ06320628 LENZ06320637 LENZ06320649 LENZ06320658 LENZ06320670 LENZ06321049 LENZ06321058 LENZ06321070 LENZ06321073 LENZ06321076 LENZ06321079 LENZ06321085 LENZ06321088 LENZ06321094 LENZ06321573 LENZ06321576 LENZ06321579 LENZ06321588 LENZ06321594
Overhead stirrer tools, anchor stirrers
Stirrers Stirring Tools
Anchor stirrers with GL thread connection.
  • Made of precision calibrated glass tube
  • With ground and polished bearing surface
  • Suitable for flange vessels