KPG stirrers with lateral blade
Supplier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
06307201 06307010 06307003 06307301 06307303 06307305 06307311 06307001 06307005 06307008 06307011 06307013 06307015 06307205 06307208 06307210 06307211 06307213 06307215 06307310 06307400
441-9670EA 154 GBP
441-9670 441-9679 441-9683 LENZ06307301 LENZ06307303 LENZ06307305 LENZ06307311 441-1000 441-1001 441-1002 441-1003 441-1004 441-1005 LENZ06307205 LENZ06307208 LENZ06307210 LENZ06307211 LENZ06307213 LENZ06307215 LENZ06307310 LENZ06307400
KPG stirrers with lateral blade
Stirrers Stirring Tools
DURAN®, borosilicate glass with PTFE paddle and paddle safety securing screw. 10 mm shaft Ø. The length of the ground and polished bearing surface for the shaft Ø is 160 mm.