Flat flange lids, centre neck, 1 angled side neck
Supplier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
06102444 06102111 06102141 06102214 06102244 06102264 06102464 06102544 06102564
LENZ06102444EA 715 GBP
LENZ06102444 LENZ06102111 LENZ06102141 LENZ06102214 LENZ06102244 LENZ06102264 LENZ06102464 LENZ06102544 LENZ06102564
Flat flange lids, centre neck, 1 angled side neck
Lids Reaction Vessel Lids
DURAN®, Borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Flange lids for reaction vessels, with laboratory flange LF.