Density bottles, adjusted
Supplier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
05630210 05630213 05630222 05630228 05630237
LENZ05630210EA 141 GBP
LENZ05630210 LENZ05630213 LENZ05630222 LENZ05630228 641-0900
Density bottles, adjusted
Specific Gravity Bottles
Borosilicate glass 3.3. DURAN®, Gay-Lussac type.
  • The density bottles (pycnometers) are individually engraved with the actual capacity, with an accuracy of 0,001 cm³
  • The top of stopper is ground and polished for ease of wiping clean
  • Stopper is size NS 10/19 with capillary
* DIN ISO 3507