Adapters, delivery, bent
Supplier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
05265001 05265002 05265003 05265004
LENZ05265001EA 41.7 GBP
LENZ05265001 201-3109 LENZ05265003 201-3110
Adapters, delivery, bent
Distillation Components Distillation Adapters
DURAN®, borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With core and jacket. Inclined at an angle of 105°. Cone with drip point.