Condensers, coil, double walled
Supplier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
05250201 05250303 05250304 05250204 05250404 05250504
LENZ05250201EA 234 GBP
LENZ05250201 LENZ05250303 201-0904 201-1664 LENZ05250404 LENZ05250504
Condensers, coil, double walled
Condensers
DURAN®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Coil condensers acc. to DIN 12593 and in addition to DIN, for condensing vapours, with cooling coil and jacket. With NS socket, NS cone and glass olives for water connection.
Order Now
SPECIFICATIONS
Environmentally Preferable