Threaded ball flanges
Supplier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
05108110 05108216 05108230
201-1163EA 267 GBP
201-1163 LENZ05108216 LENZ05108230
Threaded ball flanges
Adapters Adapters, Spherical Ground Joints
Made of stainless steel.
  • Very good chemical resistance
  • High temperature resistance
  • Sturdy
With KF socket, with thread.