Still head for steam distillation
Supplier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
05033044
LENZ05033044EA 155 GBP
LENZ05033044
Still head for steam distillation
Distillation Components Distillation Heads
DURAN®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With two cones NS 29/32 and one DIN thread GL 14.