Nitrogen receivers (schlenk tubes)
Supplier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
03370622 03370637 03370628 03370613 03370813 03370828 03370837 03370849
LENZ03370622EA 157 GBP
LENZ03370622 LENZ03370637 LENZ03370628 LENZ03370613 LENZ03370813 LENZ03370828 LENZ03370837 LENZ03370849
Nitrogen receivers (schlenk tubes)
Flasks Nitrogen Flasks
DURAN®, borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With 14/23 standard ground sockets/cones and PTFE needle-valve stopcock with tube connector.