Nitrogen receivers (schlenk Tubes)
Supplier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
03370149 03370213 03370222 03370237 03370113 03370122 03370128 03370137 03370228 03370249
LENZ03370149EA 160 GBP
LENZ03370149 LENZ03370213 LENZ03370222 LENZ03370237 201-1161 LENZ03370122 LENZ03370128 LENZ03370137 LENZ03370228 LENZ03370249
Nitrogen receivers (schlenk Tubes)
Flasks Nitrogen Flasks
DURAN®, borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With 14/23 standard ground sockets/cones and standard ground side stopcock with tube connector.