Attachable screw thread tubes
Supplier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
01300018 01300025 01300032 01300045 01300014
LENZ01300018EA 6.2 GBP
LENZ01300018 201-1606 201-1607 LENZ01300045 LENZ01300014
Attachable screw thread tubes
Adapters Adapters, Screwthread tubes
DURAN®, borosilicate glass, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
DIN screw thread GL.