Ball / flange connections, DURAN®
Supplier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
01250015 01250025 01260015 01260025
LENZ01250015EA 67.8 GBP
LENZ01250015 LENZ01250025 LENZ01260015 LENZ01260025
Ball / flange connections, DURAN®
Adapters Adapters, Spherical Ground Joints
Made from DURAN® tubing.
  • High temperature resistance
  • Very good chemical resistance
Manufactured to inch dimensions in accordance with DIN 12244 and DIN 12264.