Warming cabinets
Supplier: LEEC LTD
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
W157
LEECW157EA 0 GBP
LEECW157
Warming cabinets
Incubators Standard Incubators
They are suitable for warming bottles, bags of fluid/saline, and blankets to 37 °C or above.
  • Stainless steel construction
  • Fully sealed chamber
  • Fan assisted air circulation
  • Accurate temperature control
  • Energy efficient
  • Radius corners - easy to clean and disinfect
Warming cabinets are designed for use in hospitals in Operating Theatres, Accident and Emergency departments (A & E / casualty), and also maternity delivery suites.