Centrifugal Concentrators, Bench Top, CentriVap®
Supplier: LABCONCO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
7460030 7811030 7810033 7460035 7810030 7810031 7810032 7810034 7810035 7810036 7810037 7811035 7970030 7970035 7385030 7385035 7811031 7811036
531-0162EA 6300 GBP
531-0162 531-0168 531-0835 LBCP7460035 521-1545 521-1546 521-1547 531-0964 531-0965 531-0966 531-0967 LBCP7811035 531-0836 531-0968 521-1544 LBCP7385035 LBCP7811031 LBCP7811036
Centrifugal Concentrators, Bench Top, CentriVap®
Evaporators Centrifugal Evaporators
CentriVap® vacuum concentrators are available in a range of configurations. The CentriVap® DNA model is supplied with a built-in 34 litres/min diaphragm pump, two 177 ml glass traps and a DNA rotor (60×1,5/2,0 and 72×0,5 ml). Model with glass lid is recommended for evaporation of solvents that are not compatible with acrylic (acetone, acetonitrile, ethanol, DMSO, etc.). The acid resistant model is recommended for corrosive solvents (TFA, acetic acid, etc.).
  • Powder coated aluminium chamber; PTFE coated aluminium chamber with Hastelloy shaft for acid
More Product Information