Purifier® Cell Logic®+ Biosafety Cabinets B2
Supplier: LABCONCO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
303480011 303481151 303481131
LBCP303480011EA 0 GBP
LBCP303480011 LBCP303481151 LBCP303481131
Purifier® Cell Logic®+ Biosafety Cabinets B2
Fume Hoods and Enclosures Biological Safety Cabinets
Purifier® Logic®+ type B2 biosafety cabinets combine ergonomics, comfort and safety.
  • MyLogic OS displays cabinet status and alarms with clear, concise images and messaging to build user confidence
  • Constant Airflow Profile™ (CAP) Technology automatically maintains a precise volume of air as the filters load to protect a user and their samples
  • Ergonomic Inclination™ Technology Suite includes 10° angled sash, patented curved air grille with built-in armrest, fully accessible controls and eye-level mounted color display
  • Quiet, efficient ECM blower and energy
More Product Information