Protector® Laboratory Hoods, Premier
Supplier: LABCONCO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
100400061 100600021 100500061 100400020 100400051 100600020 100400002 100600050
LBCP100400061EA 0 GBP
LBCP100400061 LBCP100600021 LBCP100500061 LBCP100400020 LBCP100400051 LBCP100600020 LBCP100400002 LBCP100600050
Protector® Laboratory Hoods, Premier
Fume Hoods and Enclosures Chemical Fume Hoods
Protector premier hoods are suitable for general chemistry applications and also explosion-proof models are available for laboratories with potentially explosive/flammable atmospheres. Hoods are available with a built-in standard fiberglass blower, or an explosion-proof fiberglass blower, or without a blower (remote blower sold separately).
  • With glacier white powder-coated steel exterior which is durable and corrosion resistant
  • Removable front and side panels
  • Fluorescent lighting with vapor proof design
These hoods incorporate a sleek interior with aMore Product Information