Calibration thermostats, ECO
Supplier: LAUDA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
L002848 L002850
LAUDL002848EA 9310 GBP
LAUDL002848 LAUDL002850
Calibration thermostats, ECO
Circulating Thermostats Open Circulator Systems Calibration Circulators
Calibration thermostatic bath with latest microprocessor technology and integrated cooling system. Working temperature range: −25 to 200 °C
  • Safety class III for operation with flammable and non-flammable liquids. Over-temperature cut-out adjustable via menu
  • Various pump with six adjustable performance levels
  • Easy control of the flow rate between internal and external circulation during operation without contact with the bath
  • USB interface as standard
  • 2-chamber construction for constant level in the thermosetting chamber
  • Vertical adjustment of the overflow
More Product Information