Laboratory glassware washers-dryer, 910 LX Ultima
Supplier: GETINGE LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
910LX 01260100 01260285 01260293 01260301 01260276 01260284 01260292 01260300 01260277
141-0023EA 19560 GBP
141-0023 141-0287 LANC01260285 LANC01260293 LANC01260301 LANC01260276 LANC01260284 LANC01260292 LANC01260300 LANC01260277
Laboratory glassware washers-dryer, 910 LX Ultima
Glassware Washing Machines
The Lancer 910 LX offers 2 levels of cleaning with a choice of position for the upper level. This freestanding glassware washer while being contained in a small footprint, does not compromise on wash capacity. It includes a built-in drawer for the safe storage and input of chemicals. It also offers powerful forced air drying through the injectors and in the chamber to ensure complete and rapid drying of glassware both inside and out. Rack systems are common across the 810-1300 LX and 1800 LXA models enabling economical upgrade when expanding the capacity of your lab.
  • Fast cycles and
More Product Information