Jerricans, narrow neck, HDPE, with screwed stopcock
Supplier: LAMAPLAST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
3963 3961S
LAMA3963EA 52.4 GBP
LAMA3963 LAMA3961S
Jerricans, narrow neck, HDPE, with screwed stopcock
Bottles Jerricans
HDPE, natural.
  • Supplied with self sealing plastirob stopcock
  • Self sealing cap
  • Good chemical and temperature resistance
Ideal for use in food industries.