Polyethylene Heavy-Duty Graduated Buckets
Supplier: LAMAPLAST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
161 162 163 165 164
LAMA161EA 10.9 GBP
LAMA161 LAMA162 LAMA163 LAMA165 LAMA164
Polyethylene Heavy-Duty Graduated Buckets
Buckets
Made with low density polyethylene (LDPE), in white or black colour options.
  • Conical shape, litre graduations
  • With ergonomic handle
  • Food contact material approved
Buckets feature a flat bottom design to ensure total emptying and allow for easy cleaning.
Certifications: EU 10/2011.
Order Now
SPECIFICATIONS
Environmentally Preferable