Hermetic Tamper Evident Buckets, Rectangular
Supplier: LAMAPLAST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1109 1107 1105
LAMA1109EA 15.1 GBP
LAMA1109 LAMA1107 LAMA1105
Hermetic Tamper Evident Buckets, Rectangular
Buckets
Robust PP bucket with ergonomic handle. It features pressure lid with opening-off seal.
  • Rectangular shape
  • Flat bottom ensures total emptying and easy cleaning
  • Especially suitable in cosmetic, pharma, paint or dye industries
  • Food contact material approved
Hot filling possible up to 95 °C.