Potassium methyl malonate 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Potassium 3-methoxy-3-oxopropanoate, Monomethyl monopotassium malonate, Monomethyl malonate potassium salt

L20429.14 L20429.22
L20429.14EA 35.9 GBP
L20429.14 L20429.22
Potassium methyl malonate 98%
Potassium methyl malonate

Purity: 98% CAS No.: 38330-80-2 Molecular Formula: CH3O2CCH2CO2K Formula Weight: 156.18 Melting Point: ca 190° dec. MDL No.: MFCD00014021

Formula: C₄H₅KO₄
MW: 156.18 g/mol
Melting Pt: ∼ 190 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00014021
CAS Number: 38330-80-2
EINECS: 253-886-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR