(S)-(-)-1-Phenylpropyl isocyanate 95%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

L20246.03
L20246.03EA 56.2 GBP
L20246.03
(S)-(-)-1-Phenylpropyl isocyanate 95%
(S)-(-)-1-Phenylpropyl isocyanate

Purity: 95% CAS No.: 164033-12-9 Molecular Formula: C10H11NO Formula Weight: 161.20 Storage Temperature: 2°C to 8°C Refractive Index: 1.5090 Boiling Point: 105°/17mm Beilstein Reference: 6799586 Optical Rotation: -38.5°(c=1, chloroform) UN No.: UN2206 MDL No.: MFCD05664090

Formula: C₁₀H₁₁NO
MW: 161.20 g/mol
Boiling Pt: 105 °C (17 mmHg)
MDL Number: MFCD05664090
CAS Number: 164033-12-9
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR