2-Bromo-6-methylpyridine ≥98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

L20018.06 L20018.14
L20018.06EA 47.9 GBP
L20018.06 L20018.14
2-Bromo-6-methylpyridine ≥98%
2-Bromo-6-methylpyridine

Purity: 98% CAS No.: 5315-25-3 Molecular Formula: C6H6BrN Formula Weight: 172.03 Flash Point: 97°(206°F) Refractive Index: 1.5625 Boiling Point: 102-103°/20mm Beilstein Reference: 107322 MDL No.: MFCD00040743

Formula: C₆H₆BrN
Boiling Pt: 102…103 °C (20 mmHg)
Density: 1.512
Flash Pt: 97 °C (206 °F)
Storage Temperature: Ambient
MDL Number: MFCD00040743
CAS Number: 5315-25-3
EINECS: 226-173-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR