3-Amino-2-bromopyridine 97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

L19839.03 L19839.06 L19839.14
L19839.03EA 22.3 GBP
L19839.03 L19839.06 L19839.14
3-Amino-2-bromopyridine 97%
3-Amino-2-bromopyridine

Purity: 98% CAS No.: 39856-58-1 Molecular Formula: C5H5BrN2 Formula Weight: 173.01 Melting Point: 75-79° Beilstein Reference: 109830

Formula: C₅H₅BrN₂
Melting Pt: 75…79 °C
Storage Temperature: Ambient
MDL Number: MFCD00234064
CAS Number: 39856-58-1
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR