1-Hydroxyethylferrocene ≥99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 1-Ferrocenylethanol, (1-Hydroxyethyl)ferrocene, α-Hydroxyethylferrocene

L19681.03 L19681.06
L19681.03EA 14.5 GBP
L19681.03 L19681.06
1-Hydroxyethylferrocene ≥99%
1-Hydroxyethylferrocene

Purity: 99% CAS No.: 1277-49-2 Molecular Formula: C12H14FeO Formula Weight: 230.09 Flash Point: 100°(212°F) Melting Point: 77-79° MDL No.: MFCD00013752

Formula: C₁₂H₁₄FeO
MW: 221.01 g/mol
Flash Pt: >100 °C (212 °F)
MDL Number: MFCD00013752
CAS Number: 1277-49-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR