Spermine-(N,N'-bis(3-aminopropyl)-1,4-butanediamine) 97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

L19562.03 L19562.06
L19562.03EA 29.2 GBP
L19562.03 L19562.06
Spermine-(N,N'-bis(3-aminopropyl)-1,4-butanediamine) 97%
Spermine (N,N'-Bis(3-aminopropyl)-1,4-butanediamine)

Purity: 97% CAS No.: 71-44-3 Molecular Formula: C10H26N4 Formula Weight: 202.35 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Melting Point: 28-30° Boiling Point: 150°/5mm Beilstein Reference: 1751791 Merck Reference: 14,8743 UN No.: 3259 MDL No.: MFCD00008215

Formula: C₁₀H₂₆N₄
Boiling Pt: 150 °C (5 mmHg)
Melting Pt: 28…30 °C
Flash Pt: >110 °C (230 °F)
MDL Number: MFCD00008215
CAS Number: 71-44-3
EINECS: 200-754-2
ADR: 8,III
Merck Index: 14,8743

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR