4-(Trifluoromethyl)nicotinic acid ≥98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

L19553.03 L19553.MD
L19553.03EA 90.4 GBP
L19553.03 L19553.MD
4-(Trifluoromethyl)nicotinic acid ≥98%
4-(Trifluoromethyl)nicotinic acid

Purity: 98% CAS No.: 158063-66-2 Molecular Formula: C7H4F3NO2 Formula Weight: 191.11 Melting Point: 147-149° MDL No.: MFCD00082626

Formula: C₇H₄F₃NO₂
Melting Pt: 147…149 °C
Storage Temperature: Ambient
MDL Number: MFCD00082626
CAS Number: 158063-66-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR