4-Hydroxydiphenylamine 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: 4-(Phenylamino)phenol

L18945.09 L18945.18 L18945.00
L18945.09EA 49.4 GBP
L18945.09 L18945.18 L18945.00
4-Hydroxydiphenylamine 98%
4-Hydroxydiphenylamine

Purity: 98% CAS No.: 122-37-2 Molecular Formula: C12H11NO Formula Weight: 185.23 Flash Point: 209°(408°F) Odour: Odourless Boiling Point: 215-218°/12mm Beilstein Reference: 511942 MDL No.: MFCD00020142

Formula: C₁₂H₁₁NO
MW: 185.23 g/mol
Boiling Pt: 215…218 °C (12 mmHg)
Melting Pt: 66…71 °C
Flash Pt: 209 °C (408 °F)
Storage Temperature: Ambient
MDL Number: MFCD00020142
CAS Number: 122-37-2
EINECS: 204-538-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR