Indium(III) chloride, anhydrous 98+%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Indium Trichloride Chloride, Indium trichloride

L18758.06 L18758.14
L18758.06EA 48.3 GBP
L18758.06 L18758.14
Indium(III) chloride, anhydrous 98+%
Indium (III) chloride

Purity: 98+% CAS No.: 10025-82-8 Molecular Formula: InCl3 Formula Weight: 221.18 Odour: Odourless Melting Point: 586° Boiling Point: 800° Merck Reference: 14,4958 UN No.: 3260 MDL No.: MFCD00011058

Formula: Cl₃In
Boiling Pt: 800 °C
Melting Pt: 586 °C
Density: 3.460
Storage Temperature: Ambient
MDL Number: MFCD00011058
CAS Number: 10025-82-8
EINECS: 233-043-0
UN: 3260
ADR: 8,II
Merck Index: 14,4958

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR