6-Chloro-o-toluic acid ≥97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 2-Chloro-6-methylbenzoic acid

L18348.06 L18348.03
L18348.06EA 176 GBP
L18348.06 L18348.03
6-Chloro-o-toluic acid ≥97%
6-Chloro-o-toluic acid

Purity: 97% CAS No.: 21327-86-6 Molecular Formula: C8H7ClO2 Formula Weight: 170.60 Melting Point: 103-105° Beilstein Reference: 2614092

Formula: C₈H₇ClO₂
Melting Pt: 103…105 °C
Storage Temperature: Ambient
MDL Number: MFCD00045799
CAS Number: 21327-86-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR